Human mutation
Mutations in genes encoding polycomb repressive complex 2 subunits cause Weaver syndrome.
PMID 28229514
Weaver syndrome (WS) is a rare congenital overgrowth disorder caused by heterozygous mutations in EZH2 (enhancer of zeste homolog 2) or EED (embryonic ectoderm development). EZH2 and EED are core components of the polycomb repressive complex 2 (PRC2), which possesses histone methyltransferase activity and catalyzes trimethylation of histone H3 at lysine 27. Here, we analyzed eight probands with clinically suspected WS by whole-exome sequencing and identified three mutations: a 25.4-kb deletion partially involving EZH2 and CUL1 (individual 1), a missense mutation (c.707G>C, p.Arg236Thr) in EED (individual 2), and a missense mutation (c.1829A>T, p.Glu610Val) in SUZ12 (suppressor of zeste 12 homolog) (individual 3) inherited from her father (individual 4) with a mosaic mutation. SUZ12 is another component of PRC2 and germline mutations in SUZ12 have not been previously reported in humans. In vitro functional analyses demonstrated that the identified EED and SUZ12 missense mutations cause decreased trimethylation of lysine 27 of histone H3. These data indicate that loss-of-function mutations of PRC2 components are an important cause of WS.