EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
Protective effect of a polysaccharide from Anoectochilus roxburghii against carbon tetrachloride-induced acute liver injury in mice.
PMID 28229921
Abstract
Anoectochilus roxburghii (Wall.) Lindl. is traditionally used for the treatment of various types of chronic and acute hepatitis in China. Considering that Anoectochilus roxburghii polysaccharide (ARPT) is the main constituent of Anoectochilus roxburghii, the present study was designed to investigate the hepatoprotective effect of ARPT and its possible mechanism in carbon tetrachloride (CCl The hepatoprotective activity of ARPT (150, 300 and 500mg/kg) were investigated on CCl ARPT significantly decreased serum ALT and AST activities, hepatic MDA level, and markedly enhanced antioxidant enzyme (SOD, CAT and GSH-Px) activities and GSH level in hepatic tissue, in a dose-dependent manner, when compared to the model group. Histopathological observation revealed the hepatoprotective effect of ARPT against the damage. Furthermore, ARPT remarkably inhibited CYP2E1 mRNA expression, decreased NF-κB p65 expression and therefore to prevent the secretion of pro-inflammatory cytokines (TNF-α and IL-6) and chemokines (MCP-1, MIP-2 and KC), suppressed TGF-β1 expression and hepatocytes apoptosis. Moreover, ARPT could prevent DNA fragmentation based on TUNEL assay results. These findings suggested that ARPT possessed hepatoprotective effect against CCl