EMAIL THIS PAGE TO A FRIEND
International journal of biological macromolecules
Nanostructured polysaccharidic microcapsules for intracellular release of cisplatin.
PMID 28232109
Abstract
Carbohydrate polimeric microcapsules were assembled using a LbL approach onto a CaCO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
448877
Chitosan, medium molecular weight