EMAIL THIS PAGE TO A FRIEND
Food & function
Molecular mechanisms of the effects of the ethanolic extract of Muntingia calabura Linn. fruit on lipopolysaccharide-induced pro-inflammatory mediators in macrophages.
PMID 28232982
Abstract
Four flavonoids (epicatechin, rutin, diosmin and luteolin) and 11 phenolic acids (gallic acid, gentisic acid, p-hydroxybezoic acid, vanillic acid, caffeic acid, p-coumaric acid, ferulic acid, sinapic acid, syringic acid, p-anisic acid and rosmarinic acid) were determined in the ethanolic extract of M. calabura Linn. fruit gathered in Taiwan. The extract suppressed the lipopolysaccharide-stimulated expressions of inducible nitric oxide synthase and cyclooxygenase-2 as well as the productions of nitric oxide, prostaglandin E