EMAIL THIS PAGE TO A FRIEND
Journal of applied microbiology
Sensitive and specific detection of viable Mycobacterium avium subsp. paratuberculosis in raw milk by the peptide-mediated magnetic separation-phage assay.
PMID 28235155
Abstract
To validate an optimized peptide-mediated magnetic separation (PMS)-phage assay for detection of viable Mycobacterium avium subsp. paratuberculosis (MAP) in milk. Inclusivity, specificity and limit of detection 50% (LOD The optimized PMS-phage assay is the most sensitive and specific method available for the detection of viable MAP in milk. Further work is needed to streamline the PMS-phage assay, because the assay's multistep format currently makes it unsuitable for adoption by the dairy industry as a screening test. The inclusivity (ability to detect all MAP strains), specificity (ability to detect only MAP) and detection sensitivity (ability to detect low numbers of MAP) of the optimized PMS-phage assay have been comprehensively demonstrated for the first time.