EMAIL THIS PAGE TO A FRIEND
Cellular signalling
Role of Akt/PKB and PFKFB isoenzymes in the control of glycolysis, cell proliferation and protein synthesis in mitogen-stimulated thymocytes.
PMID 28235572
Abstract
Proliferating cells depend on glycolysis mainly to supply precursors for macromolecular synthesis. Fructose 2,6-bisphosphate (Fru-2,6-P
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU083501
MISSION® esiRNA, esiRNA human AKT1 (esiRNA1)