EMAIL THIS PAGE TO A FRIEND
Biomaterials
Orthogonal near-infrared upconversion co-regulated site-specific O
PMID 28235648
Abstract
Pre-existing hypoxia in tumors can result in an inadequate oxygen supply during photodynamic therapy (PDT), which in turn hampers photodynamic efficacy. To overcome this problem, we developed an orthogonal near-infrared upconversion controlled red blood cell (RBC) microcarriers to selectively deliver O
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56485
N-Hydroxysulfosuccinimide sodium salt, ≥98% (HPLC)
C4H4NNaO6S