EMAIL THIS PAGE TO A FRIEND
The Science of the total environment
Human and soil exposure during mechanical chlorpyrifos, myclobutanil and copper oxychloride application in a peach orchard in Argentina.
PMID 28237465
Abstract
The objective of this study was to measure the impact of the mechanized chlorpyrifos, copper oxychloride and myclobutanil application in a small peach orchard, on humans (operators, bystanders and residents) and on the productive soil. The mean Potential Dermal Exposure (PDE) of the workers (tractor drivers) was 30.8mL·h
Related Materials