EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
LY6/PLAUR domain containing 3 has a role in the maintenance of colorectal cancer stem-like cells.
PMID 28238780
Abstract
Colorectal carcinoma (CRC) is one of the most frequently diagnosed cancers and the leading cause of cancer-related death for both men and women. Recent studies have revealed that a small sub-population of cancer cells, termed cancer stem-like cells (CSCs)/cancer-initiating cells (CICs), are endowed with tumor-initiating ability, self-renewal ability and differentiation ability. CSCs/CICs are resistant to current therapies including chemotherapy and radiotherapy. Thus, CSCs/CICs are responsible for recurrence and metastasis, and eradication of CSCs/CICs is essential to cure cancer. In this study, we isolated CR-CSCs/CICs as sphere-cultured cells and found that a product derived from LY6/PLAUR domain containing 3 (LYPD3) is preferentially expressed in CSCs/CICs. Gene overexpression and gene knockdown experiments revealed that LYPD3 has a role in the maintenance of CR-CSCs/CICs. The findings provide a novel molecular insight into CR-CSCs/CICs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU035911 MISSION® esiRNA, esiRNA human LYPD3 (esiRNA1)