EMAIL THIS PAGE TO A FRIEND
Free radical biology & medicine
Systematic in vitro assessment of responses of roGFP2-based probes to physiologically relevant oxidant species.
PMID 28242229
Abstract
The genetically encoded probes roGFP2-Orp1 and Grx1-roGFP2 have been designed to be selectively oxidized by hydrogen peroxide (H
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N4148
S-Nitrosoglutathione, ≥97%
C10H16N4O7S