EMAIL THIS PAGE TO A FRIEND
Gigiena truda i professional'nye zabolevaniia
[Toxicologic characteristics of combined pesticides on the base polycarbacin].
PMID 2824299
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45577 Metiram, PESTANAL®, analytical standard