EMAIL THIS PAGE TO A FRIEND
Iranian journal of pharmaceutical research : IJPR
Nimodipine-Loaded Pluronic
PMID 28243263
Abstract
Nimodipine (NM), as a lipophilic calcium channel blocker indicated for the prevention and treatment of neurological disorders, suffers from an extensive first pass metabolism, resulting in low oral bioavailability. Polymeric micelles, self-assembled from amphiphilic polymers, have a core-shell structure which makes them unique nano-carriers with excellent performance as drug delivery. This investigation was aimed to develop NM-loaded polymeric micelles and evaluate their potential to cross the blood brain barrier (BBB). Micelles from Pluronics
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
122564
Mechlorethamine hydrochloride, 98%
C5H11Cl2N · HCl
P6500
Pentylenetetrazole
C6H10N4