EMAIL THIS PAGE TO A FRIEND
Phytotherapy research : PTR
Exploring the Effect of Ginsenoside Rh1 in a Sleep Deprivation-Induced Mouse Memory Impairment Model.
PMID 28244162
Abstract
Panax ginseng C.A. Meyer (Araliaceae) has been used in traditional Chinese medicine for enhancing cognition for thousands of years. Ginsenoside Rh1, a constituent of ginseng root, as with other constituents, has memory-improving effects in normal mice and scopolamine-induced amnesic mice. Sleep deprivation (SD) is associated with memory impairment through induction of oxidative stress. The present study investigated the effect of Rh1 against SD-induced cognitive impairment and attempted to define the possible mechanisms involved. Ginsenoside Rh1 (20 μmol/kg; 40 μmol/kg) and modafinil (0.42 g/kg) were administered to the mice intraperitoneally for 23 days. After 14-day SD, locomotor activity was examined using the open field test, and the object location recognition and Morris water maze tests were used to evaluate cognitive ability. The cortex and hippocampus were then dissected and homogenized, and levels and activities of antioxidant defense biomarkers were evaluated to determine the level of oxidative stress. The results revealed that Rh1 prevented cognitive impairment induced by SD, and its ability to reduce oxidative stress in cortex and hippocampus may contribute to the mechanism of action. Copyright © 2017 John Wiley & Sons, Ltd.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V3204
4-Vinylpyridine, contains 100 ppm hydroquinone as inhibitor, 95%
C7H7N