Protein and peptide letters
Towards Binding Mechanism of Cu2+ on Creatine Kinase from Pelodiscus sinensis: Molecular Dynamics Simulation Integrating Inhibition Kinetics Study.
PMID 28245755
Cu2+ is well known to play important roles in living organisms having bifacial distinction: essential microelement that is necessary for a wide range of metabolic processes but hyper-accumulation of Cu2+ can be toxic. The physiological function of Cu2+ in ectothermic animals such as Pelodiscus sinensis (Chinese soft-shelled turtle) has not been elucidated. In this study, we elucidated effect of Cu2+ on the energy producing metabolic enzyme creatine kinase (CK), which might directly affect energy metabolism and homeostasis of P. sinensis. We first conducted molecular dynamics (MD) simulations between P-CK and Cu2+ and conducted the inactivation kinetics including spectrofluorimetry study. MD simulation showed that Cu2+ blocked the binding site of the ATP cofactor, indicating that Cu2+ could directly inactivate P-CK. We prepared the muscle type of CK (P-CK) and confirmed that Cu2+ conspicuously inactivated the activity of P-CK (IC50 = 24.3 μM) and exhibited non-competitive inhibition manner with creatine and ATP in a first-order kinetic process. This result was well matched to the MD simulation results that Cu2+-induced non-competitive inactivation of P-CK. The spectrofluorimetry study revealed that Cu2+ induced tertiary structure changes in PCK accompanying with the exposure of hydrophobic surfaces. Interestingly, the addition of osmolytes (glycine, proline, and liquaemin) effectively restored activity of the Cu2+-inactivated P-CK. Our study illustrates the Cu2+-mediated unfolding of P-CK with disruption of the enzymatic function and the protective restoration role of osmolytes on P-CK inactivation. This study provides information of interest on P-CK as a metabolic enzyme of ectothermic animal in response to Cu2+ binding.