EMAIL THIS PAGE TO A FRIEND
Fish physiology and biochemistry
Purification and development of ELISAs for two forms of vitellogenin in Indian walking catfish, Clarias batrachus (L.).
PMID 28247155
Abstract
Two forms of vitellogenin (Vg: Vg1 and Vg2) were purified from the plasma of estradiol-17β (E