EMAIL THIS PAGE TO A FRIEND
Aquatic toxicology (Amsterdam, Netherlands)
Calcium mediates the cellular response of Chlamydomonas reinhardtii to the emerging aquatic pollutant Triclosan.
PMID 28249228
Abstract
The present study was aimed at investigating the role of intracellular free calcium, [Ca