EMAIL THIS PAGE TO A FRIEND
Journal of materials science. Materials in medicine
Extended pulsated drug release from PLGA-based minirods.
PMID 28251469
Abstract
The kinetics of degradation and sustained cancer drugs (paclitaxel (PT) and prodigiosin (PG)) release are presented for minirods (each with diameter of ~5 and ~6 mm thick). Drug release and degradation mechanisms were studied from solvent-casted cancer drug-based minirods under in vitro conditions in phosphate buffer solution (PBS) at a pH of 7.4. The immersed minirods were mechanically agitated at 60 revolutions per minute (rpm) under incubation at 37 °C throughout the period of the study. The kinetics of drug release was studied using ultraviolet visible spectrometry (UV-Vis). This was used to determine the amount of drug released at 535 nm for poly(lactic-co-glycolic acid) loaded with prodigiosin (PLGA-PG) samples, and at 210 nm, for paclitaxel-loaded samples (PLGA-PT). The degradation characteristics of PLGA-PG and PLGA-PT are elucidated using optical microscope as well as scanning electron microscope (SEM). Statistical analysis of drug release and degradation mechanisms of PLGA-based minirods were performed. The implications of the results are discussed for potential applications in implantable/degradable structures for multi-pulse cancer drug delivery.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
303143
3,6-Dimethyl-1,4-dioxane-2,5-dione, 99%
C6H8O4
G1796
Glycolide, ≥99%
C4H4O4