EMAIL THIS PAGE TO A FRIEND
Nature communications
Enhancing the catalytic activity of hydronium ions through constrained environments.
PMID 28252021
Abstract
The dehydration of alcohols is involved in many organic conversions but has to overcome high free-energy barriers in water. Here we demonstrate that hydronium ions confined in the nanopores of zeolite HBEA catalyse aqueous phase dehydration of cyclohexanol at a rate significantly higher than hydronium ions in water. This rate enhancement is not related to a shift in mechanism; for both cases, the dehydration of cyclohexanol occurs via an E1 mechanism with the cleavage of C
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T81108
1,3,5-Trioxane, ≥99%
C3H6O3