EMAIL THIS PAGE TO A FRIEND
Plant physiology and biochemistry : PPB
Simultaneous knock-down of six β-galactosidase genes in petunia petals prevents loss of pectic galactan but decreases petal strength.
PMID 28254702
Abstract
Galactose (Gal) is incorporated into cell wall polysaccharides as flowers open, but then is lost because of β-galactosidase activity as flowers mature and wilt. The significance of this for flower physiology resides in the role of galactan-containing polysaccharides in the cell wall, which is still largely unresolved. To investigate this, transcript accumulation of six cell wall-associated β-galactosidases was simultaneously knocked down in 'Mitchell' petunia (Petunia axillaris x (P. axillaris x P. hybrida)) flower petals. The multi-PhBGAL RNAi construct targeted three bud- and three senescence-associated β-galactosidase genes. The petals of the most down-regulated line (GA19) were significantly disrupted in galactose turnover during flower opening, and at the onset of senescence had retained 86% of their galactose compared with 20% in the controls. The Gal content of Na
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
199524
Chlorophenol Red, indicator grade
C19H12Cl2O5S