EMAIL THIS PAGE TO A FRIEND
Chemosphere
Occurrence and distribution of organophosphate esters in urban soils of the subtropical city, Guangzhou, China.
PMID 28254716
Abstract
67 soil samples from the road greenbelts, the paddy/vegetable fields, the parks, the commercial and residential areas in the subtropical city, Guangzhou, China, were collected and analyzed for 11 organophosphate esters (OPEs) and triphenylphosphine oxide (TPPO). OPEs were detected in all soil samples analyzed, which indicate that OPEs are ubiquitous environmental contaminants. The ∑
Related Materials