EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Synthesis of new photosensitive H
PMID 28256650
Abstract
A new two-electron photosensitizer, H
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
385638
Di-(2-picolyl)amine, 97%
C12H13N3