EMAIL THIS PAGE TO A FRIEND
Journal of leukocyte biology
Chemical composition and phagocyte immunomodulatory activity of
PMID 28258152
Abstract
Essential oil extracts from
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C191
Capsazepine, ≥98% (HPLC), solid
C19H21ClN2O2S