EMAIL THIS PAGE TO A FRIEND
Journal of leukocyte biology
Rationalized design of a mucosal vaccine protects against
PMID 28258153
Abstract
Pulmonary tuberculosis (TB) caused by
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0196
Resiquimod, ≥98% (HPLC)
C17H22N4O2