EMAIL THIS PAGE TO A FRIEND
Reproduction (Cambridge, England)
Maternal metabolic stress may affect oviduct gatekeeper function.
PMID 28258156
Abstract
We hypothesized that elevated non-esterified fatty acids (NEFA) modify
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
190144
Pyridinium chlorochromate, 98%
C5H5N · ClCrO3H