EMAIL THIS PAGE TO A FRIEND
Tropical animal health and production
Effect of changes in the nutritional status on the performances of growing Creole kids during an established nematode parasite infection.
PMID 28258541
Abstract
In this study, we evaluated the effect of changes in the nutritional status on the performances of growing Creole kids during an established experimental gastrointestinal nematode (GIN) infection. Eighteen 6-month-old Creole kids were distributed in two main groups infected (I) and non-infected (NI) and were placed for a period of 4 weeks on each of three diets differing in their nutritional values: (1) fresh grass (FG, 6.7 MJ/kg dry matter (DM) and 7.9% crude protein (CP)) non-supplemented, (2) FG supplemented with a commercial concentrate (CC, 12.2 MJ/kg DM and 20.6% of CP), and (3) FG supplemented with dried banana (Ban, 11.1 MJ/kg DM and 4.3% CP). The experiment was designed as a split-plot with experimental infection (I and NI) as the main plot and the diets (FG, CC, and Ban) as the subplots with three replicates. We showed a significant effect of the diet changes on the fecal egg counts. A higher dry matter intake, digestibility, and growth rate were observed with the CC diet but together with a slight but significant increase of the intensity of the GIN infection. These data suggest that the improvement of the protein nutritional status during an establish GIN infection would improve the animal performance at the expense of the mechanism involved in the control of the infection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34000
Toltrazuril, VETRANAL, analytical standard
C18H14F3N3O4S