EMAIL THIS PAGE TO A FRIEND
Molecular immunology
Characterizations of CD4-1, CD4-2 and CD8β T cell subpopulations in peripheral blood leucocytes, spleen and head kidney of Japanese flounder (Paralichthys olivaceus).
PMID 28260650
Abstract
In the previous study, antibodies against CD3 molecule have been produced and were used in labeling T cells in Japanese flounder (Paralichthys olivaceus). In this paper, CD4