EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Presence of Methicillin Resistant Staphylococcus aureus (MRSA) in sewage treatment plant.
PMID 28262305
Abstract
The presence of antibiotic resistant bacteria and antibiotic resistance genes in rural sewage treatment plants are not well reported in the literature. The aim of the present study was to study the frequency occurrence of Methicillin Resistant Staphylococcus aureus (MRSA) in a rural sewage treatment plant. This study was conducted using raw sewage as well as treated sewage from a small town sewage treatment plant in rural southeast Louisiana of USA. Results showed the presence of MRSA consistently in both raw and treated sewage. The presence of mecA gene responsible for methicillin resistance was confirmed in the raw and treated sewage water samples.