EMAIL THIS PAGE TO A FRIEND
Scientific reports
High frequency stimulation induces sonic hedgehog release from hippocampal neurons.
PMID 28262835
Abstract
Sonic hedgehog (SHH) as a secreted protein is important for neuronal development in the central nervous system (CNS). However, the mechanism about SHH release remains largely unknown. Here, we showed that SHH was expressed mainly in the synaptic vesicles of hippocampus in both young postnatal and adult rats. High, but not low, frequency stimulation, induces SHH release from the neurons. Moreover, removal of extracellular Ca