EMAIL THIS PAGE TO A FRIEND
Head & neck
Correlation between fluorodeoxyglucose hotspots on pretreatment positron emission tomography/CT and preferential sites of local relapse after chemoradiotherapy for head and neck squamous cell carcinoma.
PMID 28263422
Abstract
The potential benefits of Seventy-two consecutive patients with locally advanced HNSCC treated by RT ± chemotherapy were included in this study. All patients underwent FDG-PET/CT at initial staging (PET Nineteen patients (26%) had local relapses. Using a 40% SUVmax threshold, the initial metabolic tumor volume was significantly higher in patients with local relapses than in controlled patients (10.4 ± 8.6 vs 5.1 ± 4.9 cc; p = .002) as well as total lesion glycolysis (117.9 ± 88.6 vs 60.6 ± 80.4; p = .013). For both methods, the overlap index among A Our study does not find high overlap index values between the initial tumor and recurrence subvolumes, probably because of a suboptimal coregistration. Our results also confirm that metabolic tumor volume and total lesion glycolysis are independently correlated with recurrence-free survival in patients with HNSCC. Further larger prospective studies with FDG-PET/CT performed in the same RT position and with a validated elastic registration method are needed. © 2017 Wiley Periodicals, Inc. Head Neck 39: 1155-1165, 2017.