Journal of biomedical materials research. Part A
Inhibition of multi-species oral biofilm by bromide doped bioactive glass.
PMID 28263451
Bioactive glass is an attractive biomaterial that has shown excellent osteogenic and angiogenic effects for oral bone repairing procedures. However, anti-biofilm potential related to such biomaterial has not been completely validated, mainly against multi-species biofilms involved in early tissue infections. The aim of the present study was to evaluate the anti-biofilm effect of 58 S bioactive glass embedding calcium bromide compounds at different concentrations. Bioactive glass free or containing 5, or 10 wt % CaBr2 was synthesized by alkali sol-gel method and then characterized by physco-chemical analyses and scanning electron microscopy (SEM). Then, samples were tested by microbiological assays using optical density, real time q-PCR, and SEM. Bioactive glass particles showed accurate chemical composition and an angular shape with a bimodal size distribution ranging from 0.6 to 110 µm. The mean particle size was around 29 µm. Anti-biofilm effect was recorded for 5 wt % CaBr2 -doped bioactive glass against S. mitis, V. parvula, P. gingivais, S. gordoni, A. viscosus, F, nucleatum, P. gingivais. F. nucleatum, and P. gingivalis. Such species are involved in the biofilm structure related to infections on hard and soft tissues in the oral cavity. The incorporation of calcium bromide into bioactive glass can be a strategy to enhance the anti-biofilm potential of bioactive glasses for bone healing and infection treatment. © 2016 Wiley Periodicals, Inc. J Biomed Mater Res Part A: 105A: 1994-2003, 2017.
Related Materials
Product #
Molecular Formula
Add to Cart
Ethyltriphenylphosphonium bromide, 99%