International journal of biological macromolecules
Vitamin B12 offers neuronal cell protection by inhibiting Aβ-42 amyloid fibrillation.
PMID 28267610
Protein misfolding and aggregation has been implicated as the cause of more than 20 diseases in humans such as Alzheimer's and Parkinson's and systemic amyloidosis. Retardation of Aβ- 42 aggregation is considered as a promising and challenging strategy for developing effective therapeutics against Alzheimer's disease. Herein, we demonstrated the effect of vitamin B12 (VB) on inhibiting amyloid formation by employing ThT fluorescence assay, circular dichroism, ANS fluorescence assay, dynamic light scattering measurements and transmission electron microscopy and cell viability assay. Our results demonstrate that vitamin B12 (VB), inhibits Aβ- 42 aggregation in a concentration dependent manner. Further VB also provide protection against amyloid induced cytotoxicity in human neuronal cell line. This study points towards a promising strategy to combat Aβ- 42 aggregation and may have broader implication for targeting other neurological disorders whose distinct hallmark is also amyloid formation.