EMAIL THIS PAGE TO A FRIEND
Cancer prevention research (Philadelphia, Pa.)
Metabolic Obesity, Adipose Inflammation and Elevated Breast Aromatase in Women with Normal Body Mass Index.
PMID 28270386
Abstract
Obesity is associated with breast white adipose tissue (WAT) inflammation, elevated levels of the estrogen biosynthetic enzyme, aromatase, and systemic changes that have been linked to the pathogenesis of breast cancer. Here, we determined whether metabolic obesity, including changes in breast biology and systemic effects, occurs in a subset of women with normal body mass index (BMI). Breast WAT and fasting blood were collected from 72 women with normal BMI (<25 kg/m