EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
TNU-9 Zeolite: Aluminum Distribution and Extra-Framework Sites of Divalent Cations.
PMID 28272755
Abstract
The TNU-9 zeolite (TUN framework) is one of the most complex zeolites known. It represents a highly promising matrix for both acid and redox catalytic reactions. We present here a newly developed approach involving the use of
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
140805
1,4-Dibromobutane, 99%
C4H8Br2