EMAIL THIS PAGE TO A FRIEND
Plant physiology and biochemistry : PPB
A Glycine max homolog of NON-RACE SPECIFIC DISEASE RESISTANCE 1 (NDR1) alters defense gene expression while functioning during a resistance response to different root pathogens in different genetic backgrounds.
PMID 28273511
Abstract
A Glycine max homolog of the Arabidopsis thaliana NON-RACE SPECIFIC DISEASE RESISTANCE 1 (NDR1) coiled-coil nucleotide binding leucine rich repeat (CC-NB-LRR) defense signaling gene (Gm-NDR1-1) is expressed in root cells undergoing a defense response to the root pathogenic nematode, Heterodera glycines. Gm-NDR1-1 overexpression in the H. glycines-susceptible genotype G. max
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0662
6-Carboxyfluorescein, ~97% (HPLC)
C21H12O7