EMAIL THIS PAGE TO A FRIEND
BMC complementary and alternative medicine
Effect of microbial siderophores on mammalian non-malignant and malignant cell lines.
PMID 28274224
Abstract
Iron is a vital nutrient for all cells, and malignant cells have a higher requirement for the metal due to their rapid multiplication. Bacterial siderophores can be used to reduce free ferric ion concentration from the cellular environment. In the present study, we have evaluated effect of three siderophores - exochelin-MS, mycobactin S and deferoxamine B on the proliferation of mammalian cell lines using MTT assay. These siderophores caused a significant decrease in the viability of malignant cells, without significantly affecting non-malignant cells. Based on these results, we suggest that iron-chelation therapy could be explored as an adjunctive therapeutic option against cancer along with other therapies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M2786
3-Maleimidobenzoic acid N-hydroxysuccinimide ester, crystalline
C15H10N2O6