EMAIL THIS PAGE TO A FRIEND
Food chemistry
Detection of kanamycin and gentamicin residues in animal-derived food using IgY antibody based ic-ELISA and FPIA.
PMID 28274457
Abstract
Our aim in this study is to show that IgY antibody based immunoassays could be used to detect antibiotic residues in animal-derived food. Briefly, full antigens of gentamicin (Gent) and kanamycin (Kana) were used to immunize the laying chickens to prepare IgY antibodies. Then, these antibodies were evaluated by FPIA and ic-ELISA to detect Gent/Kana in animal-derived samples. The IC