Journal of immunology (Baltimore, Md. : 1950)
Antibody-Independent Function of Human B Cells Contributes to Antifungal T Cell Responses.
PMID 28275140
Fungal infections (e.g., Candida albicans) can manifest as serious medical illnesses, especially in the elderly and immune-compromised hosts. T cells are important for Candida control. Whether and how B cells are involved in antifungal immunity has been less clear. Although patients with agammaglobulinemia exhibit normal antifungal immunity, increased fungal infections are reported following B cell-depleting therapy, together pointing to Ab-independent roles of B cells in controlling such infections. To test how human B cells may contribute to fungal-associated human T cell responses, we developed a novel Ag-specific human T cell/B cell in vitro coculture system and found that human B cells could induce C. albicans-associated, MHC class II-restricted responses of naive T cells. Activated B cells significantly enhanced C. albicans-mediated Th1 and Th17 T cell responses, which were both strongly induced by CD80/CD86 costimulation. IL-6(+)GM-CSF(+) B cells were the major responding B cell subpopulation to C. albicans and provided efficient costimulatory signals to the T cells. In vivo B cell depletion in humans resulted in reduced C. albicans-associated T responses. Of note, the decreased Th17, but not Th1, responses could be reversed by soluble factors from B cells prior to depletion, in an IL-6-dependent manner. Taken together, our results implicate an Ab-independent cytokine-defined B cell role in human antifungal T cell responses. These findings may be particularly relevant given the prospects of chronic B cell depletion therapy use in lymphoma and autoimmune disease, as patients age and are exposed to serial combination therapies.
Related Materials
Product #
Molecular Formula
Add to Cart
Galantide, ≥90% (HPLC)