EMAIL THIS PAGE TO A FRIEND
Methods and applications in fluorescence
Rose bengal in poly(2-hydroxyethyl methacrylate) thin films: self-quenching by photoactive energy traps.
PMID 28276341
Abstract
The effect of dye concentration on the fluorescence,Φ
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
105481
1,3-Diphenylisobenzofuran, 97%
C20H14O