EMAIL THIS PAGE TO A FRIEND
Scientific reports
A potential regulatory network underlying distinct fate commitment of myogenic and adipogenic cells in skeletal muscle.
PMID 28276486
Abstract
Mechanism controlling myo-adipogenic balance in skeletal muscle is of great significance for human skeletal muscle dysfunction and myopathies as well as livestock meat quality. In the present study, two cell subpopulations with particular potency of adipogenic or myogenic differentiation were isolated from neonatal porcine longissimus dorsi using the preplate method to detect mechanisms underlying distinct fate commitment of myogenic and adipogenic cells in skeletal muscle. Both cells share a common surface expression profile of CD29