EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Synchronism in mitochondrial ROS flashes, membrane depolarization and calcium sparks in human carcinoma cells.
PMID 28279675
Abstract
Mitochondria are major producers of reactive oxygen species (ROS) in many cells including cancer cells. However, complex interrelationships between mitochondrial ROS (mitoROS), mitochondrial membrane potential (ΔΨm) and Ca