EMAIL THIS PAGE TO A FRIEND
Mutation research
Micronucleus, cell-free DNA, and plasma glycan composition in the newborns of healthy and diabetic mothers.
PMID 28283092
Abstract
Diabetes is associated with certain environmental exposures, heritable factors, and metabolic conditions of intrauterine development due to diabetes in the mother. We evaluated genomic damage, cell-free DNA, N-glycosylation of umbilical cord plasma proteins (PG), and nuclear division index (NDI) as possible prognostic biomarkers of health risk in the newborns of mothers with treated pregestational diabetes (NBDM; 22 mothers), compared these parameters with those from newborns of healthy mothers (NBHM; 89 mothers), and associated the results with the mothers' lifestyle in both groups, based on a detailed questionnaire. Genomic damage was estimated by the in vitro micronucleus (MN) assay. NDI was detected on MN slides. Glycans were analyzed by ultra-performance liquid chromatography that separates the plasma N-glycome into 46 glycan peaks. Cell-free DNA was analyzed by real-time PCR. For the association between biomarkers and individual characteristics, generalized linear/nonlinear analysis was performed. No significant difference was found between NBHM and NBDM for cell-free DNA levels. There was no association between cell-free DNA levels and lifestyle. MN frequency was significantly higher in NBDM than in NBHM (median, 0.6 vs. 0.3%, p<0.001). MN frequency and NDI were significantly associated with residence (urban vs. rural). PG differed significantly between NBHM and NBDM (p<0.001). A significant association was found between PG and increase of MN frequency (p<0.001). As both MN frequency and altered N-glycosylation are associated with cancer risk, our study indicates need for further investigations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
88071
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
C44H30N4