EMAIL THIS PAGE TO A FRIEND
The international journal of tuberculosis and lung disease : the official journal of the International Union against Tuberculosis and Lung Disease
Effect of previous treatment and sputum quality on diagnostic accuracy of Xpert
PMID 28284253
Abstract
In early studies, Xpert® MTB/RIF accurately detected culture-proven pulmonary tuberculosis (TB). Recent reports have, however, found a lower than expected specificity in previously treated TB patients. To investigate the diagnostic accuracy of Xpert in presumptive pulmonary TB patients in Southwestern Uganda. We obtained demographic and clinical information and collected three sputum samples from each patient for smear microscopy, Xpert and culture. We estimated Xpert sensitivity and specificity against culture, and stratified the analysis by previous treatment and sputum quality status. We analyzed results from 860 presumptive TB patients, including 109 (13%) with a previous history of anti-tuberculosis treatment; 205 (24%) were culture-positive. Xpert specificity was lower (91.8%, 95%CI 84.9-96.2) in previously treated than in new TB patients (97.5%, 95%CI 96.1-98.5; P = 0.01). In an adjusted analysis, patients with culture-, Xpert+ results were more likely to have been previously treated for TB (OR 8.3, 95%CI 2.1-32.0; P = 0.002), and to have mucosalivary sputum (OR 4.1, 95%CI 1.1-14.6; P = 0.03), but were less likely to self-report fever (OR 0.23, 95%CI 0.1-0.7; P = 0.008) than patients with concordant positive results. Xpert specificity was lower in previously treated patients with suspected TB. The clinical and programmatic impact of culture-, Xpert+ results requires evaluation in future studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
51362
Auramine, analytical standard
C17H21N3