EMAIL THIS PAGE TO A FRIEND
Biology of blood and marrow transplantation : journal of the American Society for Blood and Marrow Transplantation
Abnormalities of the Bone Marrow Immune Microenvironment in Patients with Prolonged Isolated Thrombocytopenia after Allogeneic Hematopoietic Stem Cell Transplantation.
PMID 28286199
Abstract
Prolonged isolated thrombocytopenia (PT) is a serious complication after allogeneic hematopoietic stem cell transplantation (allo-HSCT). Whether abnormalities of the bone marrow (BM) immune microenvironment are involved in the pathogenesis of PT remains unknown, however. Twenty patients with PT, 40 matched patients with good graft function (GGF) after allo-HSCT, and 20 healthy donors (HD) were enrolled in this nested case-control study. Th1, Th2, Tc1, Tc2, Th17, and Treg cells were analyzed by flow cytometry, and IFN-γ, IL-4, IL-17, IL-6, IL-21, and thrombopoietin levels in BM plasma were evaluated with a cytometric bead assay and ELISA. Relative to GGF patients and HD controls, PT patients had significantly higher proportions of Th1 and Tc1 cells, resulting in higher Th1/Th2 and Tc1/Tc2 ratios in the BM microenvironment. In addition, the excessive polarization of Th17 was observed in patients with PT. Changes in BM plasma cytokines were consistent with our cellular findings. These results suggest that dysregulated T cell responses in the BM microenvironment might play an important role in the pathogenesis of PT.