EMAIL THIS PAGE TO A FRIEND
Journal of animal science and biotechnology
Investigation of the immune effects of
PMID 28286647
Abstract
The health of chickens and the welfare of poultry industry are central to the efforts of addressing global food security. Therefore, it is essential to study chicken immunology to maintain and improve its health and to find novel and sustainable solutions. This paper presents a study on investigation of the effect of The 121-day-old Hubbard Hi-Y male broiler hybrids were randomly assigned to four treatment groups, three SBR supplemented groups (0.5, 1.0, and 1.5% of SBR) and one control group. Each treatment was replicated five times with six birds per replicate pen in a battery brooder. Blood was collected after 3 Results showed that mean heterophile/lymphocyte ratio increased in the SBR groups compared to the control group and the blood of the chickens showed lymphocytic depletion. The results also demonstrated that the relative weight of bursa of Fabricius and spleen in groups fed with SBR significantly decreased compared to the control group. This study also showed that the addition of SBR significantly inhibited the formation of RS of nuclei compared to some cytotoxic substances. We found that SBR supplementation should be carefully evaluated when given to poultry. The excess intake of SBR supplementation may cause immunologic inhibition and may negatively affect the development of immune organs. SBR has inhibited the formation of radial segmentation nuclei showing antimetastatic properties and also the phagocytosis of chicken heterophils.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
465119
Baicalein, 98%
C15H10O5
F2003
Flavone, ≥99%
C15H10O2
SMB00081
Wogonoside, ≥95% (LC/MS-ELSD)
C22H20O11