EMAIL THIS PAGE TO A FRIEND
Scientific reports
Novel enhancement mechanism of tyrosine hydroxylase enzymatic activity by nitric oxide through S-nitrosylation.
PMID 28287127
Abstract
Tyrosine hydroxylase (TH) is a rate-limiting step enzyme in the synthesis of catecholamines. Catecholamines function both as hormone and neurotransmitters in the peripheral and central nervous systems, therefore TH's expression and enzymatic activity is tightly regulated by various mechanisms. Several post-translational modifications have been shown to regulate TH's enzymatic activity such as phosphorylation, nitration and S-glutathionylation. While phosphorylation at N-terminal of TH can activate its enzymatic activity, nitration and S-glutathionylation can inactivate TH. In this study, we found that TH can also be S-nitrosylated by nitric oxide (NO). S-nitrosylation is a reversible modification of cysteine (cys) residue in protein and is known to be an emerging signaling mechanism mediated by NO. We found that TH can be S-nitrosylated at cys 279 and TH S-nitrosylation enhances its enzymatic activity both in vitro and in vivo. These results provide a novel mechanism of how NO can modulate TH's enzymatic activity through S-nitrosylation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N4148
S-Nitrosoglutathione, ≥97%
C10H16N4O7S