EMAIL THIS PAGE TO A FRIEND
Natural product research
Two new sesquiterpene lactone glycosides from Cnicus benedictus.
PMID 28287272
Abstract
Two new sesquiterpene lactone glycosides, namely melitensin 15-O-β-D-glucoside (1) and 11β,13-dihydrosalonitenolide 15-O-β-D-glucoside (2), along with eight known compounds (3-10) were isolated from the aerial part of Cnicus benedictus L. Their structures were elucidated from analyses of extensive spectroscopic data. Compounds 1-6 all possessed an α-methyl-γ-lactone moiety. Moreover, compound 5 exhibited moderate activity against the growth of Aspergillus fumigatus, with IC
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
410209
L-Cysteine methyl ester hydrochloride, 98%
C4H9NO2S · HCl