EMAIL THIS PAGE TO A FRIEND
Scientific reports
Restoring the impaired cardiac calcium homeostasis and cardiac function in iron overload rats by the combined deferiprone and N-acetyl cysteine.
PMID 28287621
Abstract
Intracellular calcium [Ca