EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Functional and Structural Characterization of a Thermostable Phospholipase A
PMID 28287729
Abstract
Novel phospholipase (PLA
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1263
1,2-Didodecanoyl-sn-glycero-3-phosphocholine, ≥99%, synthetic
C32H64NO8P