EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Assessment of a metal-organic framework catalyst in air cathode microbial fuel cells over time with different buffers and solutions.
PMID 28288433
Abstract
Metal-organic framework (MOF) on activated carbon (AC) enhanced the performance of cathodes but longevity needs to be considered in the presence of metal chelators or ligands, such as phosphate, present in wastewaters. MOF catalysts on AC initially produced 2.78±0.08Wm