EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Mesoscale Architecture Shapes Initiation and Richness of Spontaneous Network Activity.
PMID 28292833
Abstract
Spontaneous activity in the absence of external input, including propagating waves of activity, is a robust feature of neuronal networks